ENA018107337 Enamine

3-amino-6-ethyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03966874
  • Cas Number : 568551-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03850
Availability details for
Quantity
  • Availability
  • Add to Cart