ENA018107342 Enamine

3-(dimethylamino)benzohydrazide

  • MDL number MFCD00238588
  • Cas Number : 205927-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03855
Availability details for
Quantity
  • Availability
  • Add to Cart