ENA018107351 Enamine

ethyl 2-(carbamothioylamino)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD02140142
  • Cas Number : 105544-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03864
Availability details for
Quantity
  • Availability
  • Add to Cart