ENA018107352 Enamine

3,7-dibenzyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD00836293
  • Cas Number : 139927-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03867
Availability details for
Quantity
  • Availability
  • Add to Cart