ENA018107370 Enamine

1-(3-acetylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00609989
  • Cas Number : 95695-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03885
Availability details for
Quantity
  • Availability
  • Add to Cart