ENA018107404 Enamine

1-(4-Chloro-phenyl)-5-methoxy-3-methyl-1H-pyrazole

  • MDL number MFCD02704661
  • Cas Number : 55227-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03933
Availability details for
Quantity
  • Availability
  • Add to Cart