ENA018107460 Enamine

2-(5-Chloro-pyridin-2-yl)-4-ethoxymethylene-4H-isoquinoline-1,3-dione

  • MDL number MFCD03656265
  • Cas Number : 750607-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04002

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04002
Availability details for
Quantity
  • Availability
  • Add to Cart