ENA018107463 Enamine

4-(4-Ethoxymethylene-1,3-dioxo-3,4-dihydro-1H-isoquinolin-2-yl)-benzoic acid ethyl ester

  • MDL number MFCD03656261
  • Cas Number : 750607-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04005

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04005
Availability details for
Quantity
  • Availability
  • Add to Cart