ENA018107472 Enamine

4-(chloromethyl)-3H,4H,6H,7H,8H,9H,10H-2lambda6-[1,2,4,6]thiatriazino[4,3-a]azepine-2,2-dione

  • MDL number MFCD06654914
  • Cas Number : 885458-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04014
Availability details for
Quantity
  • Availability
  • Add to Cart