ENA018107476 Enamine

2-[2-(pyridin-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD01681558
  • Cas Number : 34272-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-147
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04018
Availability details for
Quantity
  • Availability
  • Add to Cart