ENA018107481 Enamine

6-amino-5-(2-chloroacetyl)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD04606703
  • Cas Number : 67130-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04023
Availability details for
Quantity
  • Availability
  • Add to Cart