ENA018107496 Enamine

4-(hexyloxy)-3-methoxybenzoic acid

  • MDL number MFCD03658028
  • Cas Number : 79294-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04047
Availability details for
Quantity
  • Availability
  • Add to Cart