ENA018107503 Enamine

ethyl 2-(2-chloroacetamido)-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylate

  • MDL number MFCD00436345
  • Cas Number : 203385-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04054
Availability details for
Quantity
  • Availability
  • Add to Cart