ENA018107510 Enamine

3-[(4-bromophenyl)sulfamoyl]-4-methoxybenzoic acid

  • MDL number MFCD01122302
  • Cas Number : 312274-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04061
Availability details for
Quantity
  • Availability
  • Add to Cart