ENA018107514 Enamine

4-cyclohexyl-5-(2,4-dichlorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01820438
  • Cas Number : 309280-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04066
Availability details for
Quantity
  • Availability
  • Add to Cart