ENA018107523 Enamine

3-(2-Chloro-benzoylamino)-3-phenyl-propionic acid

  • MDL number MFCD00700129
  • Cas Number : 448199-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04076
Availability details for
Quantity
  • Availability
  • Add to Cart