ENA018107534 Enamine

6-amino-1-butyl-3-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03480191
  • Cas Number : 53681-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04088
Availability details for
Quantity
  • Availability
  • Add to Cart