ENA018107535 Enamine

1-tert-butyl-3-(2-chloroacetyl)urea

  • MDL number MFCD03948988
  • Cas Number : 4791-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04089
Availability details for
Quantity
  • Availability
  • Add to Cart