ENA018107556 Enamine

2-[(3,5-dimethoxyphenyl)formamido]-3-methylbutanoic acid

  • MDL number MFCD03948993
  • Cas Number : 1397000-89-3
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04111
Availability details for
Quantity
  • Availability
  • Add to Cart