ENA018107558 Enamine

6-Amino-1-benzyl-5-(2-chloro-acetyl)-3-ethyl-1H-pyrimidine-2,4-dione

  • MDL number MFCD03964653
  • Cas Number : 568543-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04113
Availability details for
Quantity
  • Availability
  • Add to Cart