ENA018107577 Enamine

6-amino-1-butyl-5-[(2-methoxyethyl)amino]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03949033
  • Cas Number : 721414-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04135
Availability details for
Quantity
  • Availability
  • Add to Cart