ENA018107582 Enamine

2-({5-phenylthieno[2,3-d]pyrimidin-4-yl}sulfanyl)acetic acid

  • MDL number MFCD01948238
  • Cas Number : 312947-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-175
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04141
Availability details for
Quantity
  • Availability
  • Add to Cart