ENA018107595 Enamine

5-[(3-methylphenoxy)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD01710600
  • Cas Number : 66178-72-1
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04154
Availability details for
Quantity
  • Availability
  • Add to Cart