ENA018107597 Enamine

2-{[5-(Morpholine-4-sulfonyl)-2-morpholin-4-yl-phenylcarbamoyl]-methylsulfanyl}-nicotinic acid

  • MDL number MFCD03949339
  • Cas Number : 721406-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04157

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04157
Availability details for
Quantity
  • Availability
  • Add to Cart