ENA018107608 Enamine

2-{[2-oxo-3-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)propyl]sulfanyl}benzoic acid

  • MDL number MFCD06660485
  • Cas Number : 721406-44-6
  • Molecular Weight : 367.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04168
Availability details for
Quantity
  • Availability
  • Add to Cart