ENA018107609 Enamine

3-Benzothiazol-2-yl-4-(4-butoxy-phenyl)-but-3-enoic acid

  • MDL number MFCD03949336
  • Cas Number : 721406-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04169
Availability details for
Quantity
  • Availability
  • Add to Cart