ENA018107620 Enamine

1-benzyl-5-chloro-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD01136171
  • Cas Number : 100440-60-6
  • Molecular Weight : 274.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-174
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04181
Availability details for
Quantity
  • Availability
  • Add to Cart