ENA018107621 Enamine

(1-Propyl-1H-benzoimidazol-2-ylsulfanyl)-acetic acid

  • MDL number MFCD00845352
  • Cas Number : 312608-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04182

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04182
Availability details for
Quantity
  • Availability
  • Add to Cart