ENA018107624 Enamine

N1-benzyl-4-chlorobenzene-1,2-diamine

  • MDL number MFCD03965244
  • Cas Number : 39235-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04186
Availability details for
Quantity
  • Availability
  • Add to Cart