ENA018107627 Enamine

methyl 3-(4-benzyl-7-chloro-3-oxo-3,4-dihydroquinoxalin-2-yl)propanoate

  • MDL number MFCD03965264
  • Cas Number : 568544-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04189
Availability details for
Quantity
  • Availability
  • Add to Cart