ENA018107631 Enamine

1-(2-Chloro-5-trifluoromethyl-phenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02707508
  • Cas Number : 327085-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...118
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04193

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04193
Availability details for
Quantity
  • Availability
  • Add to Cart