ENA018107642 Enamine

4,5-Di-p-tolyl-1,3-dihydro-imidazole-2-thione

  • MDL number MFCD06654917
  • Cas Number : 73181-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04204

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04204
Availability details for
Quantity
  • Availability
  • Add to Cart