ENA018107643 Enamine

bis(thiophen-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD03949054
  • Cas Number : 721415-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04205
Availability details for
Quantity
  • Availability
  • Add to Cart