ENA018107647 Enamine

6-amino-5-(benzylamino)-1-butyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03949036
  • Cas Number : 721415-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04209
Availability details for
Quantity
  • Availability
  • Add to Cart