ENA018107664 Enamine

7-(4-bromophenyl)-5-hydroxy-2H-1,3-benzoxathiol-2-one

  • MDL number MFCD02575436
  • Cas Number : 327078-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04227
Availability details for
Quantity
  • Availability
  • Add to Cart