ENA018107673 Enamine

4-(2-ethoxyphenoxy)butanoic acid

  • MDL number MFCD03950714
  • Cas Number : 721416-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04236
Availability details for
Quantity
  • Availability
  • Add to Cart