ENA018107677 Enamine

N-benzyl-4-chloro-N-{[4-(hydrazinecarbonyl)phenyl]methyl}benzene-1-sulfonamide

  • MDL number MFCD03950715
  • Cas Number : 721416-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04240
Availability details for
Quantity
  • Availability
  • Add to Cart