ENA018107679 Enamine

3-Methyl-1-(3-trifluoromethyl-phenyl)-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD03949057
  • Cas Number : 721415-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 271...273
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04242
Availability details for
Quantity
  • Availability
  • Add to Cart