ENA018107689 Enamine

3-{benzyl[3-(trifluoromethyl)phenyl]sulfamoyl}-4-chlorobenzoic acid

  • MDL number MFCD03950767
  • Cas Number : 721418-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04254
Availability details for
Quantity
  • Availability
  • Add to Cart