ENA018107707 Enamine

N-(3,4-difluorophenyl)-3-oxobutanamide

  • MDL number MFCD00810466
  • Cas Number : 414872-57-4
  • Molecular Weight : 213.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3,4-difluorophenyl)-3-oxobutanamide
Inchi Code 1S/C10H9F2NO2/c1-6(14)4-10(15)13-7-2-3-8(11)9(12)5-7/h2-3,5H,4H2,1H3,(H,13,15)
Inchi Key KONWBQBDOHKZAL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04273
Availability details for
Quantity
  • Availability
  • Add to Cart