ENA018107715 Enamine

5-(2,3-Dichloro-phenyl)-furan-2-carbaldehyde

  • MDL number MFCD00579046
  • Cas Number : 106827-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04282
Availability details for
Quantity
  • Availability
  • Add to Cart