ENA018107747 Enamine

2-Hydroxy-6-m-tolylsulfamoyl-quinoline-4-carboxylic acid

  • MDL number MFCD03950771
  • Cas Number : 721418-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04321
Availability details for
Quantity
  • Availability
  • Add to Cart