ENA018107748 Enamine

3-(4-acetyl-3,5-dimethyl-1H-pyrrol-2-yl)-3-oxopropanenitrile

  • MDL number MFCD03950743
  • Cas Number : 721417-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04322
Availability details for
Quantity
  • Availability
  • Add to Cart