ENA018107750 Enamine

1-(2-Chloro-acetyl)-3-isopropyl-urea

  • MDL number MFCD03950742
  • Cas Number : 7248-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04324
Availability details for
Quantity
  • Availability
  • Add to Cart