ENA018107751 Enamine

2-[(4-methoxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-1,3-dione

  • MDL number MFCD00974750
  • Cas Number : 217493-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04325
Availability details for
Quantity
  • Availability
  • Add to Cart