ENA018107752 Enamine

2-(2-fluorobenzenesulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD03964660
  • Cas Number : 1009681-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04326
Availability details for
Quantity
  • Availability
  • Add to Cart