ENA018107758 Enamine

4,5-Di-p-tolyl-oxazol-2-ylamine

  • MDL number MFCD03964619
  • Cas Number : 726153-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04332

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04332
Availability details for
Quantity
  • Availability
  • Add to Cart