ENA018107759 Enamine

4,5-Diphenyl-oxazol-2-ylamine

  • MDL number MFCD00450541
  • Cas Number : 33119-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-04333
Availability details for
Quantity
  • Availability
  • Add to Cart