ENA018107767 Enamine

3-(1H-1,3-benzodiazol-2-yl)-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD00649220
  • Cas Number : 143361-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04342
Availability details for
Quantity
  • Availability
  • Add to Cart