ENA018107792 Enamine

3-methyl-1-(4-methylphenyl)-6-phenyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD03948174
  • Cas Number : 749216-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04369
Availability details for
Quantity
  • Availability
  • Add to Cart